missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthaldehyde, 95%

Product Code. 10051460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10051460 100 g Glass bottle 100g
10733891 500 g Glass bottle 500g
10125253 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051460 Supplier Thermo Scientific Acros Supplier No. 128091000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66-77-3 | C11H8O | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-77-3
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.18
MDL Number MFCD00004003
InChI Key SQAINHDHICKHLX-UHFFFAOYSA-N
Synonym 1-naphthaldehyde, 1-formylnaphthalene, 1-naphthalenecarboxaldehyde, 1-naphthylaldehyde, alpha-naphthal, alpha-naphthaldehyde, naphthaldehyde, .alpha.-naphthal, alpha-naphthylcarboxaldehyde, alpha-naphthylaldehyde
PubChem CID 6195
ChEBI CHEBI:52367
IUPAC Name naphthalene-1-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 1.0°C to 2.0°C
Color Brown-Yellow to Yellow
Density 1.1500g/mL
Boiling Point 160.0°C to 161.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CHO
Refractive Index 1.6500 to 1.6540
Quantity 100 g
Beilstein 07, 400
Specific Gravity 1.15
Solubility Information Solubility in water: <1g/L (20°C)
Formula Weight 156.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Naphthaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.