missing translation for 'onlineSavingsMsg'
Learn More

1-n-Propylpiperazine dihydrobromide, 98%, Thermo Scientific™

Product Code. 11443973
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443973 25 g 25g
11453973 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443973 Supplier Thermo Scientific Alfa Aesar Supplier No. A10258.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64262-23-3
Molecular Formula C7H18Br2N2
Molecular Weight (g/mol) 290.043
MDL Number MFCD00051992
InChI Key YTPQLWVHCBATKO-UHFFFAOYSA-N
Synonym 1-propylpiperazine dihydrobromide, 1-n-propylpiperazine dihydrobromide, n-propylpiperazinium dibromide, n-propylpiperazine dihydrobromide, 1-n-propylpiperazine 2hbr, piperazine, 1-propyl-, dihydrobromide, acmc-209nl5, ksc494c5l, propylpiperazine, bromide, bromide, 1-n-propyl-piperazine dihydrobromide
PubChem CID 3085288
IUPAC Name 1-propylpiperazine;dihydrobromide
SMILES CCCN1CCNCC1.Br.Br

Specifications

Melting Point ∼255°C (decomposition)
Quantity 25 g
Beilstein 3678528
Sensitivity Hygroscopic
Formula Weight 290.05
Percent Purity 98%
Chemical Name or Material 1-n-Propylpiperazine dihydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.