missing translation for 'onlineSavingsMsg'
Learn More

1-(N,N-Dimethylsulfamoyl)-1H-imidazole, TRC

Product Code. 30370543
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370543 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370543 Supplier Toronto Research Chemicals Supplier No. TRCD479315500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(N,N-Dimethylsulfamoyl)-1H-imidazole
CAS 78162-58-0
Molecular Formula C5H9N3O2S
Quantity 500 mg
Synonym 1-(N,N-Dimethylsulfamoyl)imidazole, Imidazole-1-sulfonic Acid Dimethylamide, N,N-Dimethyl-1H-imidazole-1-sulfonamide, N,N-Dimethylsulfamoyl-1H-imidazole, NSC 700551
InChI Formula InChI=1S/C5H9N3O2S/c1-7(2)11(9,10)8-4-3-6-5-8/h3-5H,1-2H3
SMILES CN(C)S(=O)(=O)n1ccnc1
IUPAC Name N,N-dimethylimidazole-1-sulfonamide
Molecular Weight (g/mol) 175.21
Formula Weight 175.0415
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.