missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyltheobromine, 98+%

Product Code. 11353167
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353167 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11353167 Supplier Thermo Scientific Alfa Aesar Supplier No. L13597.MD
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1028-33-7 | C13H20N4O2 | 264.33 g/mol

A vasodilator

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1028-33-7
Melting Point 80°C to 81°C
Molecular Formula C13H20N4O2
MDL Number MFCD00041424
Quantity 250 mg
Beilstein 270632
Merck Index 14,7127
Synonym pentifylline, 1-hexyltheobromine, 1-n-hexyltheobromine, hexyltheobromine, cosaldon, pentifyllin, cosadon, theobromine, 1-hexyl, sk 7 pharmaceutical, 1-hexyl-3,7-dimethylxanthine
InChI Key MRWQRJMESRRJJB-UHFFFAOYSA-N
SMILES CCCCCCN1C(=O)N(C)C2=C(N(C)C=N2)C1=O
IUPAC Name 1-hexyl-3,7-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular Weight (g/mol) 264.33
PubChem CID 70569
Formula Weight 264.33
Percent Purity ≥98%
Chemical Name or Material 1-n-Hexyltheobromine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.