missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-4-iodobenzene, 98%

Product Code. 11327069
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327069 1 g 1g
11337069 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327069 Supplier Thermo Scientific Alfa Aesar Supplier No. H26878.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62150-34-9 | C12H17I | 288.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62150-34-9
Molecular Formula C12H17I
Molecular Weight (g/mol) 288.17
MDL Number MFCD07777097
InChI Key QCTCAMDTUGSMJX-UHFFFAOYSA-N
Synonym 1-n-hexyl-4-iodobenzene, benzene, 1-hexyl-4-iodo, 4-hexylphenyl iodide
PubChem CID 15001645
IUPAC Name 1-hexyl-4-iodobenzene
SMILES CCCCCCC1=CC=C(I)C=C1

Specifications

Boiling Point 122°C to 125°C (3 mmHg)
Flash Point >120°C (248°F)
Refractive Index 1.553
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 288.16
Percent Purity 98%
Chemical Name or Material 1-n-Hexyl-4-iodobenzene

RUO – Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.