missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butylpyridinium chloride, 98%, Thermo Scientific Chemicals

Product Code. 11313597
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

The derivatives of 1-n-Butylpyridinium chloride is used in spent fuel reprocessing, particularly in the extraction of high-level radioactive aqueous waste from the processing of nuclear fuel. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1124-64-7
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.668
MDL Number MFCD00060047
InChI Key POKOASTYJWUQJG-UHFFFAOYSA-M
Synonym 1-butylpyridinium chloride, 1-butylpyridin-1-ium chloride, n-butylpyridinium chloride, butylpyridinium chloride, dsstox_cid_11462, dsstox_rid_78879, dsstox_gsid_31462, unii-f8m63j351a, butylpyridiniumchloride, butylpyridine, chloride
PubChem CID 2734171
IUPAC Name 1-butylpyridin-1-ium;chloride
SMILES CCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Melting Point 131°C to 132°C
Quantity 2 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 171.67
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-n-Butylpyridinium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'