missing translation for 'onlineSavingsMsg'
Learn More

1-(n-Butyl)imidazole, 99%

Product Code. 11302057
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302057 100 g 100g
11392047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302057 Supplier Thermo Scientific Alfa Aesar Supplier No. L07793.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4316-42-1 | C7H12N2 | 124.19 g/mol

1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(n-Butyl)imidazole is used in the synthesis of a heterocyclic mesomeric betaine and 1-(1-butyl-3-imidazolio)propane-3-sulfonate (BIm3S). It acts as an N-coordinated ligand. It is used in the chiral separation of amino acids and anionic pharmaceuticals by capillary electrophoresis. It is used for preparation of substituted phenyl-containing imidazolium ionic liquid.

Solubility
Soluble in Chloroform, Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4316-42-1
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00042753
InChI Key MCMFEZDRQOJKMN-UHFFFAOYSA-N
Synonym 1-butyl-1h-imidazole, 1h-imidazole, 1-butyl, n-butylimidazole, 1-n-butylimidazole, n-n-butyl imidazole, 1-n-butyl imidazole, 1-buthylimidazole, n-n-butylimidazole, imidazole, 1-butyl
PubChem CID 61347
IUPAC Name 1-butyl-1H-imidazole
SMILES CCCCN1C=CN=C1

Specifications

Density 0.948
Boiling Point 257°C
Flash Point 145°C (293°F)
Odor Amine-like
Refractive Index 1.48
Quantity 100 g
UN Number UN2690
Beilstein 109499
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 124.19
Percent Purity 99%
Chemical Name or Material 1-(n-Butyl)imidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.