missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium trifluoromethanesulfonate, 98%

Product Code. 11329715
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329715 2 g 2g
11339715 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11329715 Supplier Thermo Scientific Alfa Aesar Supplier No. L19765.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 174899-66-2 | C9H15F3N2O3S | 288.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174899-66-2
Molecular Formula C9H15F3N2O3S
Molecular Weight (g/mol) 288.285
MDL Number MFCD03427620
InChI Key FRZPYEHDSAQGAS-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium trifluoromethanesulfonate, 1-butyl-3-methylimidazolium trifluoromethansulfonate, 1-butyl-3-methylimidazolium triflate, c4mim triflate, bmim otf, acmc-1bwln, basionics™ vs 12, dsstox_cid_29088, dsstox_rid_83307, dsstox_gsid_49232
PubChem CID 2734246
IUPAC Name 1-butyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 16°C
Density 1.295
Flash Point None
Refractive Index 1.439
Quantity 2 g
Sensitivity Hygroscopic
Formula Weight 288.29
Percent Purity 98%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.