missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium nitrate, 95%, Thermo Scientific™

Product Code. 15436218
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436218 5 g 5g
15446218 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436218 Supplier Thermo Scientific Alfa Aesar Supplier No. H59803.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179075-88-8
Molecular Formula C8H15N3O3
Molecular Weight (g/mol) 201.226
MDL Number MFCD08457696
InChI Key WPHIMOZSRUCGGU-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium nitrate, 1-butyl-3-methyl-1h-imidazol-3-ium nitrate, bmim no3, c4mim nitrate, ksc171c6p, 1-butyl-3-methylimidazoliumnitrate, 1-butyl-3-methyl-imidazol-3-ium nitrate, 1-butyl-3-methylimidazolium nitrate hplc
PubChem CID 16211113
IUPAC Name 1-butyl-3-methylimidazol-3-ium;nitrate
SMILES CCCCN1C=C[N+](=C1)C.[N+](=O)([O-])[O-]

Specifications

Quantity 5 g
UN Number UN1479
Sensitivity Hygroscopic; May form a precipitate
Formula Weight 201.22
Percent Purity 95%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.