Learn More
1-n-Butyl-3-methylimidazolium hexafluorophosphate, 98+%
Descripción
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-n-Butyl-3-methylimidazolium hexafluorophosphate is used as a solvent in headspace gas chromatography and countercurrent chromatography, as mobile and stationary phases. It is widely used in organic synthesis, extraction technologies, electochemistry and biphasic organometallic catalysis.
Solubility
Miscible with dichloromethane, chloroform, ethyl acetate. Immiscible with water, diethyl ether and hexane.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Especificaciones
Especificaciones
| Melting Point | 8°C to 10°C |
| Density | 1.365 |
| Boiling Point | >350°C |
| Refractive Index | 1.4115 |
| Quantity | 100 g |
| Sensitivity | Hygroscopic |
| Merck Index | 14,1581 |
| Solubility Information | Miscible with dichloromethane,chloroform,ethyl acetate. Immiscible with water,diethyl ether and hexane. |
| Formula Weight | 284.19 |
| Percent Purity | ≥98% |
| Mostrar más |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.