missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium di-n-butyl phosphate, 96%

Product Code. 15435336
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 15435336

Brand: Thermo Scientific Alfa Aesar H59893.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 663199-28-8 | C16H33N2O4P | 348.424 g/mol

1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl-3-methylimidazolium dibutyl phosphate is employed as an imidazolium compound salt.

Solubility
Insoluble in water.

Notes
Keep away from heat and humidity. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 663199-28-8
Molecular Formula C16H33N2O4P
Molecular Weight (g/mol) 348.424
MDL Number MFCD09265043
InChI Key NTXQJRGQUZXUMU-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium dibutyl phosphate, 1-n-butyl-3-methylimidazolium di-n-butyl phosphate, 1-butyl-3-methylimidazolium dibutylphosphate
PubChem CID 16218227
IUPAC Name 1-butyl-3-methylimidazol-3-ium;dibutyl phosphate
SMILES CCCCN1C=C[N+](=C1)C.CCCCOP(=O)([O-])OCCCC

Specifications

Color Brown
Refractive Index 1.472
Quantity 50 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 348.42
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-n-Butyl-3-methylimidazolium di-n-butyl phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.