missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butoxy-2,3-difluorobenzene, 95%, Thermo Scientific™

Product Code. 15458508
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458508 25 g 25g
15448508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458508 Supplier Thermo Scientific Alfa Aesar Supplier No. H61913.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136239-66-2
Molecular Formula C10H12F2O
Molecular Weight (g/mol) 186.202
MDL Number MFCD07368738
InChI Key HYDIPEHDSUNHPA-UHFFFAOYSA-N
Synonym 3-butoxy-1,2-difluorobenzene, 2,3-difluorobutoxybenzene, benzene, 1-butoxy-2,3-difluoro, 1-butoxy-2,3-difluoro-benzene, pubchem15526, acmc-209c5k, ksc493i5l, 2,3-difluoro-1-butoxybenzene, 4-butoxy-2,3-difluorobenzene
PubChem CID 2782939
IUPAC Name 1-butoxy-2,3-difluorobenzene
SMILES CCCCOC1=C(C(=CC=C1)F)F

Specifications

Color Colorless
Quantity 25 g
Formula Weight 186.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-n-Butoxy-2,3-difluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.