missing translation for 'onlineSavingsMsg'
Learn More

1’N-Benzyl Biotin, TRC

Product Code. 30134312
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134312 100 mg 100mg
17958914 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134312 Supplier Toronto Research Chemicals Supplier No. TRCB233025100MG
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76335-62-1
Molecular Formula C17H22N2O3S
Molecular Weight (g/mol) 334.43
Synonym (3aS,4S,6aR)-Hexahydro-2-oxo-1-(phenylmethyl)-1H-thieno[3,4-d]imidazole-4-pentanoic Acid, 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-1-(phenylmethyl)-, [3aS-(3aα,4β,6aα)]-, Thieno[3,4-d]imidazoline-4-valeric acid, 1-benzyltetrahydro-2-oxo- (7CI), 5-[(3aS,4S,6aR)-3-Benzyl-2-oxohexahydrothieno[3,4-d]imidazol-4-yl]pentanoic Acid
IUPAC Name 5-[(3aR,6S,6aS)-3-benzyl-2-oxo-3a,4,6,6a-tetrahydro-1H-thieno[3,4-d]imidazol-6-yl]pentanoic acid
SMILES OC(=O)CCCC[C@@H]1SC[C@H]2[C@@H]1NC(=O)N2Cc3ccccc3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H22N2O3S/c20-15(21)9-5-4-8-14-16-13(11-23-14)19(17(22)18-16)10-12-6-2-1-3-7-12/h1-3,6-7,13-14,16H,4-5,8-11H2,(H,18,22)(H,20,21)/t13-,14-,16-/m0/s1
Formula Weight 334.1351
Chemical Name or Material 1'N-Benzyl Biotin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.