missing translation for 'onlineSavingsMsg'
Learn More

1-Myristoyl-sn-glycero-3-phosphocholine, TRC

Product Code. 30012299
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012299 1 g 1g
30011293 500 mg 500mg
17999994 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012299 Supplier Toronto Research Chemicals Supplier No. TRCM8850101G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20559-16-4
Molecular Formula C22H46NO7P
Molecular Weight (g/mol) 467.58
Synonym (7R)-,7-Dihydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphatricosan-1-aminium Inner salt 4-Oxide, (R)-,7-Dihydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphatricosan-1-aminium Inner salt 4-Oxide, 1-Myristoyl-2-hydroxy-sn-glycero-3-phosphocholine, 1-Myristoyl-L-α-phosphorylcholine, 1-Myristoyl-sn-glycero-3-phosphatidylcholine, 1-Myristoyl-sn-glycero-3-phosphocholine, 1-Tetradecanoyl-sn-glycero-3-phosphorylcholine, Coatsome MC 40H, L-α-Myristoyllysophosphatidylcholine, M-LysoPC, Myristoyl L-α-lysolecithin, Myristoyl L-α-Lysophosphatidylcholine
IUPAC Name [(2R)-2-hydroxy-3-tetradecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H46NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-22(25)28-19-21(24)20-30-31(26,27)29-18-17-23(2,3)4/h21,24H,5-20H2,1-4H3/t21-/m1/s1
Formula Weight 467.3012
Chemical Name or Material 1-Myristoyl-sn-glycero-3-phosphocholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.