Learn More
GPE Scientific™ Norell™ Spare Tube Caps for NMR Sampling Tubes
Product Code. 12872316
Click to view available options
Color:
White
Yellow
Unit Size:
Pack of 100
Product Code. 12872316
Brand: GPE Scientific™ TC5EVA1YELLOW
Only null left
Add to basket
Add to basket
Provides for safe and secure closure of NMR sampling tubes during routine applications. Norell™ Spare Tube Caps for NMR Sampling Tubes have been developed for quality and precision and are the top choice for today's scientist.
  • Manufactured from ethylene-vinyl acetate
  • Available in different colors
Specifications
Color Yellow