missing translation for 'onlineSavingsMsg'
Learn More

1-(Methylsulfonyl)piperazine, 97%

Product Code. p-7055324
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15490809

Brand: Thermo Scientific Alfa Aesar H63870.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55276-43-2 | C5H13N2O2S | 165.23 g/mol

1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Methylsulfonyl)piperazine is employed as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55276-43-2
Molecular Formula C5H13N2O2S
Molecular Weight (g/mol) 165.23
MDL Number MFCD02153225
InChI Key ZZAKLGGGMWORRT-UHFFFAOYSA-O
Synonym 1-methylsulfonyl piperazine, 1-methanesulfonylpiperazine, 1-methanesulfonyl-piperazine, 1-methylsulfonyl-piperazine, 1-methylsulphonyl piperazine, methylsulfonyl piperazine, piperazine, 1-methylsulfonyl, 1-n-methanesulfonylpiperazine, 4-methylsulfonylpiperazine, n-mesylpiperazine
PubChem CID 709161
IUPAC Name 1-methylsulfonylpiperazine
SMILES CS(=O)(=O)N1CC[NH2+]CC1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 164.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Methylsulfonyl)piperazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.