missing translation for 'onlineSavingsMsg'
Learn More

1-Methylsulfonyl-1H-benzotriazole, 97%

Product Code. 15490148
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490148 25 g 25g
15470148 1 g 1g
15480148 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490148 Supplier Thermo Scientific Alfa Aesar Supplier No. H53386.14
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 37073-15-7 | C7H7N3O2S | 197.21 g/mol

1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylsulfonyl-1H-benzotriazole is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37073-15-7
Molecular Formula C7H7N3O2S
Molecular Weight (g/mol) 197.21
MDL Number MFCD00595638
InChI Key OMMZSDGGKVRGGP-UHFFFAOYSA-N
Synonym 1-methylsulfonyl-1h-benzotriazole, 1-methylsulfonyl-1h-benzo d 1,2,3 triazole, 1-methanesulfonyl-1h-benzotriazole, 1-methanesulfonyl-1,2,3-benzotriazole, acmc-20ap9j, cambridge id 5284919, methylsulfonyl benzotriazole, 1-methylsulfonyl benzotriazole, 1-methylsulphonyl-1h-benzotriazole
PubChem CID 792044
SMILES CS(=O)(=O)N1N=NC2=CC=CC=C12

Specifications

Melting Point 106°C to 108°C (decomposition)
Quantity 25 g
IUPAC Name 1-methylsulfonylbenzotriazole
Formula Weight 197.21
Percent Purity 97%
Chemical Name or Material 1-Methylsulfonyl-1H-benzotriazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.