missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole-2-carboxaldehyde, 98%

Product Code. 11469093
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469093 250 g 250g
11449093 10 g 10g
11459093 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469093 Supplier Thermo Scientific Alfa Aesar Supplier No. A11163.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-58-1 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-58-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00003087
InChI Key OUKQTRFCDKSEPL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
PubChem CID 14504
IUPAC Name 1-methylpyrrole-2-carbaldehyde
SMILES CN1C=CC=C1C=O

Specifications

Density 1.074
Boiling Point 88°C to 90°C (22 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.561
Quantity 250 g
Beilstein 107811
Sensitivity Air and light sensitive
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrole-2-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.