missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole-2-carbonyl chloride, 95%

Product Code. 11364995
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364995 1 g 1g
11374995 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364995 Supplier Thermo Scientific Alfa Aesar Supplier No. L18431.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26214-68-6 | C6H6ClNO | 143.57 g/mol

It is applied in the preparation of 5-(Pyrrolylcarbonyl)- and 5-(Imidazolylcarbonyl)pyrimidines and use also in the novel fluorescent, minor groove and sequence specific recognition element: design, synthesis, and DNA binding properties of p-Anisylbenzimidazole-imidazole/pyrrole-containing polyamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the preparation of 5-(Pyrrolylcarbonyl)- and 5-(Imidazolylcarbonyl)pyrimidines and use also in the novel fluorescent, minor groove and sequence specific recognition element: design, synthesis, and DNA binding properties of p-Anisylbenzimidazole-imidazole/pyrrole-containing polyamides.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store away from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26214-68-6
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD02094021
InChI Key JOYDZQJINHJNPM-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbonyl chloride, 1h-pyrrole-2-carbonyl chloride, 1-methyl, 1-methyl-1h-pyrrole-2-carboxylic acid chloride, acmc-1chjw, 1-methyl-2-pyrroyl chloride, n-methyl-2-pyrroloyl chloride, n-methylpyrrolecarbonyl chloride, n-methylpyrrole carbonyl chloride, 1-methylpyrrole-2-carbonylchloride, 1-methylpyrrole-2-carboyl chloride
PubChem CID 2795092
IUPAC Name 1-methylpyrrole-2-carbonyl chloride
SMILES CN1C=CC=C1C(=O)Cl

Specifications

Boiling Point 72°C to 74°C (10 mmHg)
Refractive Index 1.57
Quantity 1 g
UN Number UN3265
Beilstein 472022
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 143.57
Percent Purity 95%
Chemical Name or Material 1-Methylpyrrole-2-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.