missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpsilocin, Tocris Bioscience™
Potent and selective 5-HT2C agonist
£163.79 - £596.49
Chemical Identifiers
CAS | 1465-16-3 |
---|---|
Molecular Formula | C13H18N2O |
Molecular Weight (g/mol) | 218.3 |
InChI Key | MZZRFEIDRWKTKJ-UHFFFAOYSA-N |
Synonym | 1-methylpsilocin, 3-2-dimethylamino ethyl-1-methylindol-4-ol, 3-2-dimethylamino-ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, d0r8vt |
PubChem CID | 44404883 |
IUPAC Name | 3-[2-(dimethylamino)ethyl]-1-methylindol-4-ol |
SMILES | CN1C=C(C2=C1C=CC=C2O)CCN(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18015882
|
Tocris Bioscience
3017/10 |
10 mg |
£163.79
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18084122
|
Tocris Bioscience
3017/50 |
50 mg |
£596.49
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective 5-HT2C agonist (IC50 values are 12 and 633 nM for 5-HT2C and 5-HT2A receptors respectively). Displays high affinity for the 5-HT2B receptor (Ki = 38 nM) but acts as an inverse agonist. Active in vivo; inhibits scratching in a mouse model of OCD.Chemical Identifiers
1465-16-3 | |
218.3 | |
1-methylpsilocin, 3-2-dimethylamino ethyl-1-methylindol-4-ol, 3-2-dimethylamino-ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, d0r8vt | |
3-[2-(dimethylamino)ethyl]-1-methylindol-4-ol |
C13H18N2O | |
MZZRFEIDRWKTKJ-UHFFFAOYSA-N | |
44404883 | |
CN1C=C(C2=C1C=CC=C2O)CCN(C)C |
Specifications
1465-16-3 | |
1-methylpsilocin, 3-2-dimethylamino ethyl-1-methylindol-4-ol, 3-2-dimethylamino-ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, d0r8vt | |
CN1C=C(C2=C1C=CC=C2O)CCN(C)C | |
218.3 | |
218.29 | |
1-Methylpsilocin |
C13H18N2O | |
MZZRFEIDRWKTKJ-UHFFFAOYSA-N | |
3-[2-(dimethylamino)ethyl]-1-methylindol-4-ol | |
44404883 | |
>99% |
Safety and Handling
Recommended Storage : Store at +4°C