missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine, 99%

Product Code. 11332136
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332136 10 g 10g
11342136 100 g 100g
11352136 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332136 Supplier Thermo Scientific Alfa Aesar Supplier No. L03398.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-67-5 | C6H14ClN | 135.64 g/mol

1-Methylpiperidine is a reactant for sp3 C-H Bond activation with ruthenium(II) catalysts and C(3)-alkylation of cyclic amines, One-pot synthesis of Z-cinnamic acids. It is used in the synthesis of Unsymmetrical ureas, Antibacterial imidazolium, pyrrolidinium, and piperidinium salts, C1-C16 segment of goniodomin A via palladium-catalyzed organostannane thioester coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpiperidine is a reactant for sp3 C-H Bond activation with ruthenium(II) catalysts and C(3)-alkylation of cyclic amines, One-pot synthesis of Z-cinnamic acids. It is used in the synthesis of Unsymmetrical ureas, Antibacterial imidazolium, pyrrolidinium, and piperidinium salts, C1-C16 segment of goniodomin A via palladium-catalyzed organostannane thioester coupling.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with.acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-67-5
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00006491
InChI Key QDUXDCXILAPLAG-UHFFFAOYSA-N
Synonym n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
PubChem CID 12291
IUPAC Name 1-methylpiperidine
SMILES [H+].[Cl-].CN1CCCCC1

Specifications

Melting Point -50°C
Density 0.816
pH 12
Boiling Point 105°C to 107°C
Flash Point 3°C (37°F)
Odor Amine-like
Refractive Index 1.4378
Quantity 10 g
UN Number UN2399
Beilstein 1073
Solubility Information Fully miscible in water.
Formula Weight 99.18
Percent Purity 99%
Chemical Name or Material 1-Methylpiperidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.