missing translation for 'onlineSavingsMsg'
Learn More

(1-Methylpiperid-3-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10164104
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10164104 1 g Amber Glass Bottle 1g
10516154 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10164104 Supplier Thermo Scientific Maybridge Supplier No. CC66113DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14613-37-7
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.219
MDL Number MFCD06738902
InChI Key KEDTYNCWGSIWBK-UHFFFAOYSA-N
Synonym 1-methylpiperidin-3-yl methanamine, 3-aminomethyl-1-methylpiperidine, 1-methyl-3-aminomethyl piperidine, 3-piperidinemethanamine, 1-methyl, c-1-methyl-piperidin-3-yl-methylamine, 1-methyl-3-aminomethyl-piperidine, 1-methyl-3-piperidinemethanamine, 1-1-methylpiperidin-3-yl methanamine, 1-methylpiperid-3-yl methylamine
PubChem CID 13675353
IUPAC Name (1-methylpiperidin-3-yl)methanamine
SMILES CN1CCCC(C1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methylpiperid-3-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.