missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, TRC

Product Code. 30481817
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481817 10 g 10g
30484007 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481817 Supplier Toronto Research Chemicals Supplier No. TRCM32649510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylpiperazine
CAS 109-01-3
Molecular Formula C5 H12 N2
Quantity 10 g
Synonym 1-Methylpiperazine, Cyclizine Hydrochloride Imp. A (EP), Imatinib Mesilate Imp. G (Pharmeuropa), Piperazine, 1-methyl- (8CI, 9CI, ACI), 1-Methylpiperazine (ACI), 1-Methyl-1,4-piperazine, 1-Methyltetrahydro-1,4-diazine, N-Methylpiperazine, N-Methylpiperazine, NSC 30195, NSC 30675
InChI Formula InChI=1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3
SMILES CN1CCNCC1
IUPAC Name 1-methylpiperazine
Molecular Weight (g/mol) 100.16
Formula Weight 100.1
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.