missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, TRC

Product Code. 30480471
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480471 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480471 Supplier Toronto Research Chemicals Supplier No. TRCM32649550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylpiperazine
CAS 109-01-3
Molecular Formula C5 H12 N2
Quantity 50 g
Synonym 1-Methylpiperazine, Cyclizine Hydrochloride Imp. A (EP), Imatinib Mesilate Imp. G (Pharmeuropa), Piperazine, 1-methyl- (8CI, 9CI, ACI), 1-Methylpiperazine (ACI), 1-Methyl-1,4-piperazine, 1-Methyltetrahydro-1,4-diazine, N-Methylpiperazine, N-Methylpiperazine, NSC 30195, NSC 30675
InChI Formula InChI=1S/C5H12N2/c1-7-4-2-6-3-5-7/h6H,2-5H2,1H3
SMILES CN1CCNCC1
IUPAC Name 1-methylpiperazine
Molecular Weight (g/mol) 100.16
Formula Weight 100.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.