missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine dihydrochloride, 98%, Thermo Scientific™

Product Code. 15431838
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431838 5 g 5g
15441838 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431838 Supplier Thermo Scientific Alfa Aesar Supplier No. H55536.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34352-59-5
Molecular Formula C5H14Cl2N2
Molecular Weight (g/mol) 173.081
MDL Number MFCD00012755
InChI Key AILFRWRYZZVJTL-UHFFFAOYSA-N
Synonym 1-methylpiperazine dihydrochloride, methylpiperazine, chloride, chloride, piperazine, 1-methyl-dihydrochloride, pubchem15033, methylpiperazine dihydrochloride, n-methylpiperazine dihydrochloride, piperazine, 1-methyl-, dihydrochloride
PubChem CID 2734715
IUPAC Name 1-methylpiperazine;dihydrochloride
SMILES CN1CCNCC1.Cl.Cl

Specifications

Melting Point 246°C to 250°C
Color White
Quantity 5 g
Formula Weight 173.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Methylpiperazine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.