missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, 98+%

Product Code. 11449802
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449802 2500 mL 2500mL
11429802 100 mL 100mL
11439802 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449802 Supplier Thermo Scientific Alfa Aesar Supplier No. A10837.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-01-3 | C5H12N2 | 100.165 g/mol

1-Methylpiperazine is used in the preparation of 2-(4-methyl-1-piperazinylmethyl)acrylophenone. It is involved in the preparation of 1-(4-methoxy-phenyl)-4-methyl-piperazine by reacting with 1-chloro-4-methoxy-benzene. It acts as an intermediate in the synthesis of active pharmaceutical ingredients like ofloxacin, rifampicin, clozapine, sildenafil, trifluoperazine and zopiclone. Further, it is used as a mimic template in the preparation of molecularly imprinted microspheres. In addition to this, it is used in the generation of difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpiperazine is used in the preparation of 2-(4-methyl-1-piperazinylmethyl)acrylophenone. It is involved in the preparation of 1-(4-methoxy-phenyl)-4-methyl-piperazine by reacting with 1-chloro-4-methoxy-benzene. It acts as an intermediate in the synthesis of active pharmaceutical ingredients like ofloxacin, rifampicin, clozapine, sildenafil, trifluoperazine and zopiclone. Further, it is used as a mimic template in the preparation of molecularly imprinted microspheres. In addition to this, it is used in the generation of difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

Solubility
Miscible with water and methanol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-01-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00005966
InChI Key PVOAHINGSUIXLS-UHFFFAOYSA-N
Synonym n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
PubChem CID 53167
IUPAC Name 1-methylpiperazine
SMILES CN1CCNCC1

Specifications

Melting Point -6°C
Density 0.903
Boiling Point 137°C to 139°C
Flash Point 34°C (93°F)
Odor Amine-like
Refractive Index 1.4655
Quantity 2500 mL
UN Number UN2734
Beilstein 102724
Sensitivity Air Sensitive
Solubility Information Miscible with water and methanol.
Formula Weight 100.17
Percent Purity ≥98%
Chemical Name or Material 1-Methylpiperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.