missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081039
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
109-01-3
C5H12N2
100.165
MFCD00005966
PVOAHINGSUIXLS-UHFFFAOYSA-N
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
53167
1-methylpiperazine
CN1CCNCC1
This item is not returnable. View return policy
109-01-3
C5H12N2
100.165
MFCD00005966
PVOAHINGSUIXLS-UHFFFAOYSA-N
n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
53167
1-methylpiperazine
CN1CCNCC1

1-Methylpiperazine is used in the preparation of 2-(4-methyl-1-piperazinylmethyl)acrylophenone. It is involved in the preparation of 1-(4-methoxy-phenyl)-4-methyl-piperazine by reacting with 1-chloro-4-methoxy-benzene. It acts as an intermediate in the synthesis of active pharmaceutical ingredients like ofloxacin, rifampicin, clozapine, sildenafil, trifluoperazine and zopiclone. Further, it is used as a mimic template in the preparation of molecularly imprinted microspheres. In addition to this, it is used in the generation of difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpiperazine is used in the preparation of 2-(4-methyl-1-piperazinylmethyl)acrylophenone. It is involved in the preparation of 1-(4-methoxy-phenyl)-4-methyl-piperazine by reacting with 1-chloro-4-methoxy-benzene. It acts as an intermediate in the synthesis of active pharmaceutical ingredients like ofloxacin, rifampicin, clozapine, sildenafil, trifluoperazine and zopiclone. Further, it is used as a mimic template in the preparation of molecularly imprinted microspheres. In addition to this, it is used in the generation of difunctional strong anion-exchange stationary phase from a 1,4-diazacyclohexane derivative.

Solubility
Miscible with water and methanol.

Notes
Hygroscopic. Moisture sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY
Melting Point -6°C
Density 0.903
Boiling Point 137°C to 139°C
Flash Point 34°C (93°F)
Odor Amine-like
Refractive Index 1.4655
Quantity 100 mL
UN Number UN2734
Beilstein 102724
Sensitivity Air Sensitive
Solubility Information Miscible with water and methanol.
Formula Weight 100.17
Percent Purity ≥98%
Chemical Name or Material 1-Methylpiperazine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'