missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnicotinamide Chloride, TRC

Product Code. 30478899
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478899 Supplier Toronto Research Chemicals Supplier No. TRCM3209951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylnicotinamide chloride
CAS 1005-24-9
Molecular Formula C7 H9 N2 O . Cl
Quantity 1 g
Synonym Pyridinium','3-(aminocarbonyl)-1-methyl-','chloride (1:1), 3-Carbamoyl-1-methylpyridinium chloride (6CI,7CI), Pyridinium','3-(aminocarbonyl)-1-methyl-','chloride (9CI), Pyridinium','3-carbamoyl-1-methyl-','chloride (8CI), 1-Methyl-3-carbamoylpyridinium chloride, 1-Methylnicotinamide chloride, 3-(Aminocarbonyl)-1-methylpyridinium chloride, 3-Carbamyl-1-methylpyridinium chloride, N-Methyl-3-carboxamidepyridinium chloride, N-Methylnicotinamide chloride, N1-Methylnicotinamide chloride, Nicotinamide methochloride, Trigonellamide chloride
InChI Formula InChI=1S/C7H8N2O.ClH/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H
SMILES [Cl-].C[n+]1cccc(c1)C(=O)N
IUPAC Name 1-methylpyridin-1-ium-3-carboxamide;chloride
Molecular Weight (g/mol) 172.61
Formula Weight 172.0403
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.