missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene, 96%

Product Code. 10338793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10338793 2.5 kg Glass bottle 2.5kg
10468280 100 g Glass bottle 100g
10021170 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10338793 Supplier Thermo Scientific Acros Supplier No. 127160025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-12-0 | C11H10 | 142.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-12-0
Molecular Formula C11H10
Molecular Weight (g/mol) 142.2
MDL Number MFCD00004034
InChI Key QPUYECUOLPXSFR-UHFFFAOYSA-N
Synonym alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
PubChem CID 7002
ChEBI CHEBI:50717
IUPAC Name 1-methylnaphthalene
SMILES CC1=CC=CC2=CC=CC=C12

Specifications

CAS Min % >97%
Melting Point -22.0°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CH3
Refractive Index 1.6130 to 1.6160
Quantity 2.5 kg
Beilstein 05, 566
Specific Gravity 1.02
Solubility Information Solubility in water: 26mg/l (25°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 142.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Methylnaphthalene, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methylnaphthalene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to an approved waste d.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.