missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene, 96%

Product Code. 11478226
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478226 500 g 500g
11434160 25 g 25g
11468226 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478226 Supplier Thermo Scientific Alfa Aesar Supplier No. A15327.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 90-12-0 | C11H10 | 142.201 g/mol

1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

Solubility
Miscible with water, alcohol, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-12-0
Molecular Formula C11H10
Molecular Weight (g/mol) 142.201
MDL Number MFCD00004034
InChI Key QPUYECUOLPXSFR-UHFFFAOYSA-N
Synonym alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
PubChem CID 7002
ChEBI CHEBI:50717
IUPAC Name 1-methylnaphthalene
SMILES CC1=CC=CC2=CC=CC=C12

Spezifikation

Melting Point -22°C
Density 1.024
Boiling Point 240°C to 243°C
Flash Point 82°C (179°F)
Refractive Index 1.615
Quantity 500 g
UN Number UN3082
Beilstein 506793
Solubility Information Miscible with water,alcohol,ethanol,ether and benzene.
Formula Weight 142.2
Percent Purity 96%
Chemical Name or Material 1-Methylnaphthalene
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.