missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindoline-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10193074
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193074 1 g Amber Glass Bottle 1g
10440574 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193074 Supplier Thermo Scientific Maybridge Supplier No. CC58704DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 60082-02-2
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD03208513
InChI Key OZQGVTARHLDSTI-UHFFFAOYSA-N
Synonym 1-methylindoline-5-carbaldehyde, 1-methyl-2,3-dihydro-1h-indole-5-carbaldehyde, 1-methylindoline-5-carboxaldehyde, 1-methyl-5-indolinecarbaldehyde, 1-methyl-5-formylindoline, 5-formyl-1-methylindoline, 1-methyl-5-indolinecarbaldehyde #, 2,3-dihydro-1-methyl-1h-indole-5-carboxaldehyde, 1h-indole-5-carboxaldehyde, 2,3-dihydro-1-methyl
PubChem CID 595181
IUPAC Name 1-methyl-2,3-dihydroindole-5-carbaldehyde
SMILES CN1CCC2=C1C=CC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 1-Methylindoline-5-carbaldehyde
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1-Methylindoline-5-carbaldehye
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.