missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole, TRC

Product Code. 30465663
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. Quantity unitSize
30465663 25 g 25g
1 options
This item is not returnable. View return policy

Product Code. 30465663

Brand: Toronto Research Chemicals TRCM31356025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylindole
CAS 603-76-9
Molecular Formula C9H9N
Quantity 25 g
Synonym 1-Methyl-1H-indole, N-Methylindole, NSC 212534
InChI Formula InChI=1S/C9H9N/c1-10-7-6-8-4-2-3-5-9(8)10/h2-7H,1H3
SMILES Cn1ccc2ccccc12
IUPAC Name 1-methylindole
Molecular Weight (g/mol) 131.18
Formula Weight 131.0735
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.