missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number p-7058264
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
19012-03-4
C10H9NO
159.19
MFCD00014570
KXYBYRKRRGSZCX-UHFFFAOYSA-N
1-methyl-1h-indole-3-carbaldehyde, 1-methylindole-3-carboxaldehyde, 1-methyl-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methylindole, 1-methyl-3-formylindole, n-methyl-3-formylindole, indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methyl-1h-indole, n-methyl-3-indolecarboxaldehyde
87894
1-methylindole-3-carbaldehyde
CN1C=C(C=O)C2=CC=CC=C12
This item is not returnable. View return policy
19012-03-4
C10H9NO
159.19
MFCD00014570
KXYBYRKRRGSZCX-UHFFFAOYSA-N
1-methyl-1h-indole-3-carbaldehyde, 1-methylindole-3-carboxaldehyde, 1-methyl-indole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methylindole, 1-methyl-3-formylindole, n-methyl-3-formylindole, indole-3-carboxaldehyde, 1-methyl, 3-formyl-1-methyl-1h-indole, n-methyl-3-indolecarboxaldehyde
87894
1-methylindole-3-carbaldehyde
CN1C=C(C=O)C2=CC=CC=C12

1-Methylindole-3-carboxaldehyde is used as a reactant for preparation of nitroolefins and β-nitroalcohols via microwave- or ultrasound-assisted Henry reactions, reactant for synthesis of quinolinones via three-component Ugi reaction, reactant for synthesis of α-ketoamides as inhibitors of Dengue virus protease with antiviral activity in cell-culture, reactant for preparation of thiazolopyrimidinones as inhibitors of Bcl-2 proteins, reactant for preparation of vinylindoles via Peterson olefination or olefination with Nysted reagent, reactant for preparation of indolyl alkenes from microwave-enhanced Knoevenagel condensation as antibacterial agents 1-Methylindole-3-carboxaldehyde may be used in the synthesis of (Z)-3-(1-methyl-1H-indol-3-yl)-2-(thiophen-3-yl)acrylonitrile, via base-catalyzed condensation with thiophene-3-acetonitrile. It was also used in the preparation of monomer, required for the synthesis of poly(3-vinyl-1-methylindole).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindole-3-carboxaldehyde is used as a reactant for preparation of nitroolefins and β-nitroalcohols via microwave- or ultrasound-assisted Henry reactions, reactant for synthesis of quinolinones via three-component Ugi reaction, reactant for synthesis of α-ketoamides as inhibitors of Dengue virus protease with antiviral activity in cell-culture, reactant for preparation of thiazolopyrimidinones as inhibitors of Bcl-2 proteins, reactant for preparation of vinylindoles via Peterson olefination or olefination with Nysted reagent, reactant for preparation of indolyl alkenes from microwave-enhanced Knoevenagel condensation as antibacterial agents 1-Methylindole-3-carboxaldehyde may be used in the synthesis of (Z)-3-(1-methyl-1H-indol-3-yl)-2-(thiophen-3-yl)acrylonitrile, via base-catalyzed condensation with thiophene-3-acetonitrile. It was also used in the preparation of monomer, required for the synthesis of poly(3-vinyl-1-methylindole).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY
Melting Point 67°C to 71°C
Quantity 1 g
Beilstein 121302
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 159.19
Percent Purity ≥98%
Chemical Name or Material 1-Methylindole-3-carboxaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'