missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-2-boronic acid, 95%

Product Code. 15409807
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15409807

Brand: Thermo Scientific Alfa Aesar H53028.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 191162-40-0 | C9H10BNO2 | 174.99 g/mol

Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for Suzuki-Miyaura coupling. It ia an active Pharmaceutical ingredient hence used as pharmaceutical intermediates. Also used as rawv material in organic synthesis.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191162-40-0
Molecular Formula C9H10BNO2
Molecular Weight (g/mol) 174.99
MDL Number MFCD01114668
InChI Key CBPBJUTWVXLSER-UHFFFAOYSA-N
Synonym 1-methyl-1h-indol-2-yl boronic acid, n-methylindole-2-boronic acid, 2-borono-1-methyl-1h-indole, 1-methylindol-2-yl boronic acid, 1-methyl-1h-indole-2-boronic acid, 1-methyl-1h-indol-2-yl-2-boronic acid, boronic acid,b-1-methyl-1h-indol-2-yl, boronic acid, 1-methyl-1h-indol-2-yl, acmc-209eur, n-methyl-indoleboronic acid
PubChem CID 22733820
SMILES CN1C(=CC2=CC=CC=C12)B(O)O

Specifications

Melting Point 108°C to 112°C
Quantity 250 mg
IUPAC Name (1-methyl-1H-indol-2-yl)boronic acid
Formula Weight 174.99
Percent Purity 95%
Chemical Name or Material 1-Methylindole-2-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.