missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindazole-3-carboxylic Acid (Granisetron Impurity D), TRC

Product Code. 30138040
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138040 1 g 1g
30135484 50 mg 50mg
17942495 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138040 Supplier Toronto Research Chemicals Supplier No. TRCM3134551G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50890-83-0
Molecular Formula C9 H8 N2 O2
Molecular Weight (g/mol) 176.17
Synonym 1-Methyl-1H-indazole-3-carboxylic acid, Granisetron Hydrochloride Imp. D (EP), 1H-Indazole-3-carboxylic acid, 1-methyl-, 1-Methylindazole-3-carboxylic acid, 3-Carboxy-1-methylindazole
IUPAC Name 1-methylindazole-3-carboxylic acid
SMILES Cn1nc(C(=O)O)c2ccccc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H8N2O2/c1-11-7-5-3-2-4-6(7)8(10-11)9(12)13/h2-5H,1H3,(H,12,13)
Formula Weight 176.0586
Percent Purity >95%
Chemical Name or Material 1-Methyl-1H-indazole-3-carboxylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.