missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindazole-3-carboxylic acid, 97%

Product Code. 11357069
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357069 5 g 5g
11347069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357069 Supplier Thermo Scientific Alfa Aesar Supplier No. H26879.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 50890-83-0 | C9H8N2O2 | 176.175 g/mol

1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylindazole-3-carboxylic acid is used as important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical adjuvant, Granisetron Impurity D.

Solubility
Insoluble in water. Soluble in DMSO and Methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50890-83-0
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00272569
InChI Key OVVDFORZEGKEJM-UHFFFAOYSA-N
Synonym 1-methyl-1h-indazole-3-carboxylic acid, n-methyl-indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylic acid, 1h-indazole-3-carboxylic acid, 1-methyl, 1-methyl-indazole-3-carboxylic acid, n-methylindazole-3-carboxylic acid, 1-methylindozole-3-carboxylic acid, n-methyl-indozole-3-carboxylic acid, n-methyl-1h indazole-3-carboxylic acid, 1-methyl-3-indazolecarboxylicacid
PubChem CID 689105
IUPAC Name 1-methylindazole-3-carboxylic acid
SMILES CN1C2=CC=CC=C2C(=N1)C(=O)O

Specifications

Melting Point ∽218°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water. Soluble in DMSO and Methanol.
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material 1-Methylindazole-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.