missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1-Methylimidazole, for biochemistry, anhydrous, AcroSeal™

Product Code. 16338247 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.50 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16338247 2.5 L 2.50 L
16328247 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16338247 Supplier Thermo Scientific Chemicals Supplier No. 465290025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-47-7 | C4H6N2 | 82.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-47-7
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005292
InChI Key MCTWTZJPVLRJOU-UHFFFAOYSA-N
IUPAC Name 1-methyl-1H-imidazole
SMILES CN1C=CN=C1

Specifications

Melting Point -2.0°C
Density 1.0300 g/mL
Boiling Point 198.0°C
Flash Point 92°C
Refractive Index 1.4955 to 1.4975
Quantity 2.5 L
Specific Gravity 1.03
Solubility Information Solubility aq. soln.: 145 g/L (25°C). Other solubilities: soluble in organic solvents
Formula Weight 82.12
Grade Biochemistry
Chemical Name or Material 1-Methylimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.