missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopropanecarboxylic acid, 98%

Product Code. 10674955 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10674955

Brand: Thermo Scientific Acros 194960010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6914-76-7 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6914-76-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001290
InChI Key DIZKLZKLNKQFGB-UHFFFAOYSA-N
Synonym 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa
PubChem CID 81326
IUPAC Name 1-methylcyclopropane-1-carboxylic acid
SMILES CC1(CC1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34.0°C to 36.0°C
Boiling Point 183.0°C to 185.0°C
Flash Point 84°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C3H4CO2H
Quantity 1 g
Beilstein 09, II, 5
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 1-Methylcyclopropanecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1-Methylcyclopropanecarboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.