missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentene, 98%

Product Code. 11423098
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423098 5 g 5g
11433098 25 g 25g
11443098 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423098 Supplier Thermo Scientific Alfa Aesar Supplier No. A18816.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 693-89-0 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-89-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00001397
InChI Key ATQUFXWBVZUTKO-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
PubChem CID 12746
IUPAC Name 1-methylcyclopentene
SMILES CC1=CCCC1

Specifications

Melting Point -142°C
Density 0.779
Boiling Point 73°C to 75°C
Flash Point −17°C (1°F)
Refractive Index 1.431
Quantity 5 g
UN Number UN3295
Beilstein 1900640
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 1-Methylcyclopentene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methylcyclopentene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: 1-Methylcyclopentene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.