missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentanol, 98%

Product Code. 11476446
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11476446

Brand: Thermo Scientific Alfa Aesar B24744.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1462-03-9 | C6H12O | 100.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1462-03-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001364
InChI Key CAKWRXVKWGUISE-UHFFFAOYSA-N
PubChem CID 73830
IUPAC Name 1-methylcyclopentan-1-ol
SMILES CC1(CCCC1)O

Specifications

Melting Point 36°C to 37°C
Density 0.904
Boiling Point 135°C to 136°C
Flash Point 40°C (105°F)
Quantity 100 g
UN Number UN1325
Synonym 1-methylcyclopentanol, 1-methyl cyclopentanol, cyclopentanol, 1-methyl, 1-methyl-cyclopentanol, 1-hydroxy-1-methylcyclopentane, cyclopentanol, methyl, 1-methyl-1-cyclopentanol, 1-methylcyclopentanolc, cyclopentanol,1-methyl
Formula Weight 100.16
Percent Purity 98%
Chemical Name or Material 1-Methylcyclopentanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.