missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclohexanecarboxylic acid, 99%

Product Code. 11454160
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454160 50 g 50g
11420317 5 g 5g
11430317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454160 Supplier Thermo Scientific Alfa Aesar Supplier No. B21713.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1123-25-7 | C8H14O2 | 142.198 g/mol

1-Methyl-1-cyclohexanecarboxylic acid was used as internal standard during the determination of valproic acid metabolites. Also been used as an anticonvulsant drug and causes maturation of murine neuroblastoma cells in vitro. Used in the synthesis of p38 map kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-25-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00001463
InChI Key REHQLKUNRPCYEW-UHFFFAOYSA-N
Synonym 1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference
PubChem CID 70744
IUPAC Name 1-methylcyclohexane-1-carboxylic acid
SMILES CC1(CCCCC1)C(=O)O

Specifications

Melting Point 36°C to 39°C
Boiling Point 236°C
Flash Point 100°C (212°F)
Quantity 50 g
Beilstein 1859220
Solubility Information Soluble in chloroform and methanol.
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material 1-Methylcyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.