missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcycloheptanol, 98%, Thermo Scientific Chemicals

Product Code. 11424317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424317 5 g 5g
11434317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424317 Supplier Thermo Scientific Alfa Aesar Supplier No. B20393.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Methylcycloheptanol, is used as an pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylcycloheptanol, is used as an pharmaceutical and chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3761-94-2
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00021672
InChI Key XFFKAYOHINCUNU-UHFFFAOYSA-N
Synonym 1-methylcycloheptanol, cycloheptanol, 1-methyl, 1-methylcycloheptanol-1, 1-methyl-cycloheptanol, methylcycloheptanol-1, 1-methyl cycloheptanol, cycloheptanol,1-methyl
PubChem CID 77376
IUPAC Name 1-methylcycloheptan-1-ol
SMILES CC1(CCCCCC1)O

Specifications

Refractive Index 1.472
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Methylcycloheptanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.