missing translation for 'onlineSavingsMsg'
Learn More

1-Methylbenzimidazole, 97%

Product Code. 11477146
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477146 25 g 25g
11467146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477146 Supplier Thermo Scientific Alfa Aesar Supplier No. B25472.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1632-83-3 | C8H8N2 | 132.17 g/mol

1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylbenzimidazole is the suitable reagent used as electrolyte additive for the dye-sensitized solar cells and solid-state dye-sensitized solar cells.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1632-83-3
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00192275
InChI Key FGYADSCZTQOAFK-UHFFFAOYSA-N
Synonym 1-methyl-1h-benzimidazole, 1h-benzimidazole, 1-methyl, methylbenzimidazole, unii-12lw89n19y, 1-methyl-1h-benzo d imidazole, 1h-benzimidazole, methyl, 1-methyl-1,3-benzodiazole, 1-methyl-1h-1,3-benzodiazole, pubchem7613, 1-methylbenzimidazole
PubChem CID 95890
IUPAC Name 1-methylbenzimidazole
SMILES CN1C=NC2=CC=CC=C12

Specifications

Melting Point 59°C to 62°C
Boiling Point 154°C (12 mmHg)
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 132.17
Percent Purity 99%
Chemical Name or Material 1-Methylbenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.