missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 1-Methyl-D-tryptophan
Description
1-Methyl-D-tryptophan is an indoleamine 2,3-dioxygenase (IDO) inhibitor (IC50 = 7 μM); disrupts tryptophan catabolism. Enhances the antitumor and antiviral immunoresponses of CD8+ T-cells in vitro. Reduces tumor volume in mice with xenografts overexpressing IDO.
Specifications
Specifications
| CAS | 110117-83-4 |
| Synonym | 1MT |
| Quantity | 50 mg |
| Target | Other Oxidase/Oxygenase Compounds |
| Molecular Formula | C12H14N2O2 |
| Formula Weight | 218.25 |
| Purity | >97% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?