missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-tributylstannanyl-1H-imidazole, 97%, Thermo Scientific™

Product Code. 10730754
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10730754 250 mg 250mg
10321393 1 g 1g
10492634 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10730754 Supplier Thermo Scientific Maybridge Supplier No. CC23641CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 147716-03-8
Molecular Formula C16H32N2Sn
Molecular Weight (g/mol) 371.156
InChI Key OGYWKJKAIAEDQX-UHFFFAOYSA-N
Synonym 1-methyl-5-tributylstannyl-1h-imidazole, 1-methyl-5-tributylstannanyl-1h-imidazole, 1-methyl-5-tributylstannyl imidazole, tributyl-3-methylimidazol-4-yl stannane, 1h-imidazole,1-methyl-5-tributylstannyl, 1h-imidazole, 1-methyl-5-tributylstannyl, pubchem22530, acmc-1cabs, 1-methyl-5-tributylstannylimidazole, 5-tributylstannyl-1-methylimidazole
PubChem CID 10915651
IUPAC Name tributyl-(3-methylimidazol-4-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CN1C

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 371.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-tributylstannanyl-1H-imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.