missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific™

Product Code. 15438087
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438087 1 g 1g
15499215 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438087 Supplier Thermo Scientific Alfa Aesar Supplier No. H51405.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Methyl-5-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147716-03-8
Molecular Formula C16H32N2Sn
Molecular Weight (g/mol) 371.156
MDL Number MFCD01319031
InChI Key OGYWKJKAIAEDQX-UHFFFAOYSA-N
PubChem CID 10915651
IUPAC Name tributyl-(3-methylimidazol-4-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CN=CN1C

Specifications

Quantity 1 g
UN Number UN2788
Synonym 1-methyl-5-tributylstannyl-1h-imidazole, 1-methyl-5-tributylstannanyl-1h-imidazole, 1-methyl-5-tributylstannyl imidazole, tributyl-3-methylimidazol-4-yl stannane, 1h-imidazole,1-methyl-5-tributylstannyl, 1h-imidazole, 1-methyl-5-tributylstannyl, pubchem22530, acmc-1cabs, 1-methyl-5-tributylstannylimidazole, 5-tributylstannyl-1-methylimidazole
Solubility Information Sparingly soluble in water.
Formula Weight 371.15
Percent Purity ≥90%
Chemical Name or Material 1-Methyl-5-(tri-n-butylstannyl)imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.