missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-thien-2-yl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10428904
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10428904 5 g 5g
11350647 250 mg 250mg
10153314 250 mg 250mg
10011054 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10428904 Supplier Thermo Scientific Maybridge Supplier No. CC47101DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 869901-15-5
Molecular Formula C9H8N2O2S
Molecular Weight (g/mol) 208.235
MDL Number MFCD08271952
InChI Key URCWVEMABYSGJA-UHFFFAOYSA-N
Synonym 1-methyl-5-thien-2-yl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-thiophen-2-yl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-2-thienyl, 1-methyl-5-thiophen-2-yl pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl, 1-methyl-5-2-thienyl-1h-pyrazole-3-carboxylic acid, 1-methyl-5-2-thienyl pyrazole-3-carboxylic acid, 3-carboxy-1-methyl-5-thien-2-yl-1h-pyrazole
PubChem CID 7537648
IUPAC Name 1-methyl-5-thiophen-2-ylpyrazole-3-carboxylic acid
SMILES CN1C(=CC(=N1)C(=O)O)C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 208.24
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-thien-2-yl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.