missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-5-phenyl-1h-pyrazole-3-carbaldehyde, 97%, Thermo Scientific™

Product Code. 11340637
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11340637 250 mg Amber Glass Bottle 250mg
10602285 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11340637 Supplier Thermo Scientific Maybridge Supplier No. CC46504CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 124344-94-1
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
MDL Number MFCD08271935
InChI Key AGEKFZHCMVDIFA-UHFFFAOYSA-N
Synonym 1-methyl-5-phenyl-1h-pyrazole-3-carbaldehyde, 3-formyl-1-methyl-5-phenyl-1h-pyrazole, 1-methyl-5-phenylpyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,1-methyl-5-phenyl, acmc-20e3zm, 3-formyl-1-methyl-1h-pyrazol-5-yl benzene
PubChem CID 14354446
IUPAC Name 1-methyl-5-phenylpyrazole-3-carbaldehyde
SMILES CN1C(=CC(=N1)C=O)C2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-3-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.