missing translation for 'onlineSavingsMsg'
Learn More

1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline, 97%, Thermo Scientific™

Product Code. 10622285
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-43-1
Molecular Formula C15H22BNO2
Molecular Weight (g/mol) 259.16
MDL Number MFCD09879913
InChI Key CZYIGZLFKRAXMV-UHFFFAOYSA-N
Synonym 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indoline, 1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole, 1-methylindoline-5-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-1-methylindolin-5-yl-1,3,2-dioxaborolane, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1h-indole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydroindole
PubChem CID 24229591
IUPAC Name 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole
SMILES CN1CCC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 259.15
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'