missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-4-piperidnyl)methanamine, 97%, Thermo Scientific™

Product Code. 10265844
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265844 5 g Amber Glass Bottle 5g
10511283 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265844 Supplier Thermo Scientific Maybridge Supplier No. MO01153DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7149-42-0
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD05022430
InChI Key AGTPSAZJSOQXHJ-UHFFFAOYSA-N
Synonym 1-methylpiperidin-4-yl methanamine, 1-methyl-piperidin-4-methylamine, 1-methyl-4-piperidinyl methanamine, 4-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-4-yl methanamine, 1-methyl-4-aminomethyl piperidine, 4-aminomethyl-1-methylpiperidine, c-1-methyl-piperidin-4-yl-methylamine, 1-methylpiperidine-4-methylamine
PubChem CID 81574
IUPAC Name (1-methylpiperidin-4-yl)methanamine
SMILES CN1CCC(CN)CC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methyl-4-piperidinyl)methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.